Anti-Claudin 3 Rabbit Polyclonal Antibody
USBIC5838-27-100
- Antibody type:Primary
- Antigen name:Claudin 3
- Antigen symbol:CLDN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:10mM PBS, pH 7.6, 0,2% BSA and 15mM sodium azide, before the addition of glycerol to 40%.
- Storage temperature:–20 °C
- Concentration:~0,1 mg/ml
- Immunogen:A synthetic peptide derived from the C-terminus of mouse Claudin 3. Cellular Localization: Cell membrane and cytoplasmic. MW of antigen: 22-24kD.
- Purification:purified by immunoaffinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended dilution: Immunohistology: 1:50
Type: Primary
Antigen: Claudin 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat,