Anti-ASyO2 Mouse Monoclonal Antibody [clone: 51.24]
Catalog # AGRIAS132717
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:ASYO2
- Clonality:Monoclonal
- Clone:51.24
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Spinacia oleracea,Synechocystis pcc 6803,A. thaliana
- Cross adsorption:No
- Amino acid number:131 to 140
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:synthetic peptide derived from human alpha-synuclein Glu131–Ala140
- Tested applications:Dot Blot
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ASYO2
Clonality: Monoclonal
Clone: 51.24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: A. thaliana, Spinacia oleracea, Synechocystis pcc 6803