Order Entry
Export
ContactUsLinkComponent
Anti-Lhcb2-P Rabbit Polyclonal Antibody
Anti-Lhcb2-P Rabbit Polyclonal Antibody
Catalog # AGRIAS132705
Supplier:  Agrisera
undefined
Anti-Lhcb2-P Rabbit Polyclonal Antibody
Catalog # AGRIAS132705
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    LHCB2-P
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    A. thaliana
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Modification:
    phosphorylated
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    KLH-conjugated synthetic peptide: RRT*VKSTPQS, where T* indicates phospho-Thr
  • Size:
    25 µg
  • Pk:
    25 µG

Specifications

About this item

Type: Primary
Antigen: LHCB2-P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana