Anti-Mitogen-activated protein kinase 18 Rabbit Polyclonal Antibody
AGRIAS132673
: Agrisera
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q9ZVP5
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from known Arabidopsis thaliana kinase 18 sequence
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: Mitogen-activated Protein kinase 18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana