Anti-RPS12 Rabbit Polyclonal Antibody
Catalog # AGRIAS122114
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:RPS12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant full length RPS12 of Chlamydomonas reinhardtii, P14149, expressed in E.coli
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RPS12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: full length
Host: Rabbit
Isotype:
Reactivity: Human