Anti-S14 Rabbit Polyclonal Antibody
Catalog # AGRIAS122111
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:S14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Cylindrospermopsis raciborskii cs-505,Synechocystis sp. pcc 6803
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from a C-terminal of Arabidopsis thaliana Q9SIH0
- Size:100 µl
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: S14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: cylindrospermopsis raciborskii cs-505, Synechocystis sp. pcc6803