Anti-DCL3 Rabbit Polyclonal Antibody
Catalog # AGRIAS122103
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:DCL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zea mays,Thermosynechococcus elongatus,A. thaliana,Solanum lycopersicum
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana DCL3 sequence, Q9LXW7 , At3g43920
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: DCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Solanum lycopersicum, Thermosynechococcus elongatus, Zea mays