Order Entry
Export
ContactUsLinkComponent
 
Anti-IRT1 Rabbit Polyclonal Antibody
  AGRIAS111780
 :  Agrisera
Anti-IRT1 Rabbit Polyclonal Antibody
  AGRIAS111780
 :  Agrisera

 

  • Antibody type:
    Primary
  • Antigen symbol:
    IRT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Chicken
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    KLH-conjugated synthetic peptide derived from Arabidopsis thaliana IRT1 sequence, Q38856, At4g19690
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: IRT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken