Anti-AMY Rabbit Polyclonal Antibody
AGRIAS10712
: Agrisera
- Antibody type:Primary
- Antigen name:alpha Amylase
- Antigen symbol:AMY
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:S. tuberosum
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from known Oryza sativa P17654
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: AMY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: S. tuberosum