Order Entry
Export
ContactUsLinkComponent
Anti-Deg5 Rabbit Polyclonal Antibody
Anti-Deg5 Rabbit Polyclonal Antibody
Catalog # AGRIAS10703
Supplier:  Agrisera
Anti-Deg5 Rabbit Polyclonal Antibody
Catalog # AGRIAS10703
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    DEG5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Citrullus vulgaris,
    A. thaliana,
    Physcomitrella patens
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    recombinant mature Deg5 of Arabidopsis thaliana Q9SEL7 (At4g18370) with chloroplast and thylakoid-transfer domains excluded, overexpressed in E.coli
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: DEG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Citrullus vulgaris, Physcomitrella patens