Anti-RA Rabbit Polyclonal Antibody
Catalog # AGRIAS10700
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:RA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Oryza sativa,Camelina sativa,Thiodictyon sp. cad16,Gossypium hirsutum,Zea mays,Populus balsamifera,A. thaliana,N. tabacum,Gossypium barbadense,Thellungiella salsuginea,Soy bean
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:purified, recombinant Rubisco activase from Gossypium hirsutum Q9AXG1
- Purification:purified
- Size:100 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Camelina sativa, Glycine max, Gossypium barbadense, Gossypium hirsutum, N. tabacum, Oryza sativa, Populus balsamifera, Thiodictyon sp. cad16, Thellungiella salsuginea, Zea mays