Anti-PDC Rabbit Polyclonal Antibody
AGRIAS10691
: Agrisera
- Antibody type:Primary
- Antigen symbol:PDC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Oryza sativa,Spinacia oleracea,Brassica juncea,Zea mays,Solanum tuberosum,Pisum sativum,A. thaliana,Leianthus annuus,N. tabacum
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated peptide derived from available PDC sequences including Arabidopsis thaliana
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: PDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Brassica juncea, Leianthus annuus, N. tabacum, Oryza sativa, Pisum sativum, Solanum tuberosum, Spinacia oleracea, Zea mays