Order Entry
Export
ContactUsLinkComponent
Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # AGRIAS101285
Supplier:  Agrisera
Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # AGRIAS101285
Supplier:  Agrisera

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Hamster,
    Rabbit,
    Horse,
    Sheep,
    Goat,
    Chicken,
    Human,
    Bovine,
    Guinea pig,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Storage temperature:
    +2°C to +8 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Purified Mouse IgG, whole molecule
  • Purification:
    purified
  • Size:
    2 mg
  • Pk:
    2 mg

Specifications

About this item

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Donkey
Isotype:
Reactivity: Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit, Rat, Sheep