Anti-IgG F(c) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # AGRIAS101273
Supplier: Agrisera
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG F(c)
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Goat,Human
- Cross adsorption:No
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:Purified Mouse IgG, Fc fragment
- Purification:purified
- Size:2 mg
- Pk:2 mg
Specifications
About this item
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Goat, Human