Anti-IgG (H+L) Rabbit Polyclonal Antibody
AGRIAS101265
: Agrisera
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rabbit,Goat,Human,Mouse
- Cross adsorption:No
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:Purified Goat IgG, whole molecule
- Purification:affinity-purified
- Size:2 mg
- Pk:2 mg
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Goat, Human, Mouse, Rabbit