Anti-Alpha-mannosidase Rabbit Polyclonal Antibody
Catalog # AGRIAS09452
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:alpha Mannosidase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Canavalia ensiformis
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:native alpha-mannosidase purified from jack beans
- Purification:purified
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type: Primary
Antigen: alpha Mannosidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Canavalia ensiformis