Order Entry
Export
ContactUsLinkComponent
Anti-trans-zeatin riboside Rabbit Polyclonal Antibody
Anti-trans-zeatin riboside Rabbit Polyclonal Antibody
Catalog # AGRIAS09414
Supplier:  Agrisera
Anti-trans-zeatin riboside Rabbit Polyclonal Antibody
Catalog # AGRIAS09414
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    trans-zeatin riboside
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    BSA-conjugated, via ribose, trans-zeatin riboside
  • Tested applications:
    Radio Immunoassay
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Trans-zeatin riboside
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: