Anti-Pex14p Rabbit Polyclonal Antibody
AGRIAS08372
: Agrisera
- Antibody type:Primary
- Antigen symbol:PEX14P
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zea mays,Hordeum vulgare
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant, soluble N-terminal domain of Arabidopsis thaliana Pex14p (Q9FE40, At5g62810) that mediates PEX5 and PEX19 binding. The transmembrane and coiled-coil domains of PEX14 were replaced with a dual StrepII-His6 tag.
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: PEX14P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hordeum vulgare, Zea mays