Anti-HSP70 Rabbit Polyclonal Antibody
Catalog # AGRIAS08371
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:HSP70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide conserved in known higher plant HSC70 proteins including three isoforms of Arabidopsis thaliana HSC70-1 (NP_ 001119156.1), HSC70-2 (NP_195869.1) and HSC70-3 (NP_187555.1) as well as heat shock inducible Hsp70 of Arabidopsis thaliana AT3g12580/T2E22_110 and At1g16030 and AT3g12580/T2E22_110
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana