Anti-DnaK2 Rabbit Polyclonal Antibody
AGRIAS08350
: Agrisera
- Antibody type:Primary
- Antigen symbol:DNAK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Chlamydomonas reinhardtii
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:full length recombinant DnaK2 protein derived from Synechocystis PCC 6803 DnaK2 protein sequence P22358
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: DNAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: full length
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii