Anti-GLN1/GLN2 Rabbit Polyclonal Antibody
Catalog # AGRIAS08295
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:GLN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:GLN2
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from a wide range of available sequences including all isoforms of Arabidopsis thaliana GLN1-1,1-2,1-3 and 1-4, (At5g37600, At1g66200, At3g17820, At5g16570)
- Size:200 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GLN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana