Anti-Ycf3 Rabbit Polyclonal Antibody
AGRIAS07273
: Agrisera
- Antibody type:Primary
- Antigen symbol:YCF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Chlamydomonas reinhardtii,Cyanobacteria
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:full length recombinant ycf3 protein of Chlamydomonas reinhardtii O20031, as described in Boudreau et al. 1997
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: YCF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: full length
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii, Cyanobacteria