Anti-DnaK Rabbit Polyclonal Antibody
Catalog # AGRIAS07270
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:DNAK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Multiple
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:A8JIK8
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombiant DNAK of Chlamydomonas reinhardtii,
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: DNAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Many Species