Anti-FeSOD Rabbit Polyclonal Antibody
Catalog # AGRIAS06125
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:FESOD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:overexpressed Chlamydomonas reinhardtii thioredoxine fusion protein A8IGH1, FeSOD excised from a gel piece
- Size:200 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: FESOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human