Anti-HSP70/HSC70 Rabbit Polyclonal Antibody
AGRIAS05083A
: Agrisera
- Antibody type:Primary
- Antigen symbol:HSP70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Rainbow trout
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:HSC70
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide conserved across all known sequences of HSP70 P08107 and HSC70 proteins P11142
- Purification:affinity-purified
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rainbow Trout