Anti-VIM Mouse Monoclonal Antibody [clone: UMAB159]
Catalog # ORIGUM800054
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Vimentin
- Antigen symbol:VIM
- Clonality:Monoclonal
- Clone:UMAB159
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Amino acid number:210 to 466
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:53.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 210-466 of human VIM (NP_003371) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:30 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:200
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: UMAB159
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human