Order Entry
Export
ContactUsLinkComponent
 
Anti-MSMB Mouse Monoclonal Antibody [clone: UMAB147]
  ORIGUM800044
 :  OriGene
Anti-MSMB Mouse Monoclonal Antibody [clone: UMAB147]
  ORIGUM800044
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MSMB
  • Clonality:
    Monoclonal
  • Clone:
    UMAB147
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Purified prostate specific antigen (MSMB) from human seminal fluid
  • Purification:
    purified by affinity chromatography
  • Size:
    30 μl
  • Pk:
    100 µl

 

 

Recommended Dilutions: Immunochemistry: 1:200

Type: Primary
Antigen: MSMB
Clonality: Monoclonal
Clone: UMAB147
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human