Anti-TOP2A Mouse Monoclonal Antibody [clone: UMAB146]
ORIGUM800043
: OriGene
- Antibody type:Primary
- Antigen symbol:TOP2A
- Clonality:Monoclonal
- Clone:UMAB146
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:1100 to 1531
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1100-1531 of human TOP2A(NP_001058) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:30 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:200; Immunochemistry: 1:200
Type: Primary
Antigen: TOP2A
Clonality: Monoclonal
Clone: UMAB146
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human