Anti-NBN Mouse Monoclonal Antibody [clone: UMAB99]
ORIGUM800029
: OriGene
- Antibody type:Primary
- Antigen symbol:NBN
- Clonality:Monoclonal
- Clone:UMAB99
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human
- Cross adsorption:No
- Form:Liquid
- Amino acid number:461 to 754
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:84.8 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 461-754 of human NBN (NP_002476) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:30 μl
- Pk:100 µl
Recommended Dilutions: Immunochemistry: 1:100; Immunofluorescence: 1:100
Type: Primary
Antigen: NBN
Clonality: Monoclonal
Clone: UMAB99
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey