Order Entry
Export
ContactUsLinkComponent
 
Anti-NME1 Mouse Monoclonal Antibody [clone: UMAB92]
Catalog # ORIGUM800023
Supplier:  OriGene
Anti-NME1 Mouse Monoclonal Antibody [clone: UMAB92]
Catalog # ORIGUM800023
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    NME1
  • Clonality:
    Monoclonal
  • Clone:
    UMAB92
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Dog,
    Rat
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    19.5 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human NME1 (NP_937818) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    30 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Immunochemistry: 1:100; Immunofluorescence: 1:100

Type: Primary
Antigen: NME1
Clonality: Monoclonal
Clone: UMAB92
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Rat