Anti-DICER1 Mouse Monoclonal Antibody [clone: 2B1]
Catalog # ORIGTA801636
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:DICER1
- Clonality:Monoclonal
- Clone:2B1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Amino acid number:1666 to 1922
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:218.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1666-1922 of human DICER1 (NP_085124) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:150
Type: Primary
Antigen: DICER1
Clonality: Monoclonal
Clone: 2B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human