Anti-KRT19 Mouse Monoclonal Antibody [clone: 8H7]
Catalog # ORIGTA804001
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:KRT19
- Clonality:Monoclonal
- Clone:8H7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:240 to 390
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:43.9 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 240-390 of human KRT19 (NP_002267) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150
Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: 8H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human