Anti-MCM2 Mouse Monoclonal Antibody [clone: 1H2]
ORIGTA805504
: OriGene
- Antibody type:Primary
- Antigen symbol:MCM2
- Clonality:Monoclonal
- Clone:1H2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:101.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human MCM2 (NP_004517) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: MCM2
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human