Anti-CD1C Mouse Monoclonal Antibody [clone: UMAB46]
Catalog # ORIGUM500042
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CD1C
- Clonality:Monoclonal
- Clone:UMAB46
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:35.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human CD1C(NP_001756) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:100
Type: Primary
Antigen: CD1C
Clonality: Monoclonal
Clone: UMAB46
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human