Anti-CDH4 Mouse Monoclonal Antibody [clone: 4F5]
ORIGTA804735
: OriGene
- Antibody type:Primary
- Antigen symbol:CDH4
- Clonality:Monoclonal
- Clone:4F5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:170 to 428
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:98.3 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 170-428 of human CDH4 (NP_001785) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: CDH4
Clonality: Monoclonal
Clone: 4F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human