Anti-PTGS2 Mouse Monoclonal Antibody [clone: 9H10]
Catalog # ORIGTA805365
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:PTGS2
- Clonality:Monoclonal
- Clone:9H10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:18 to 260
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:67.2 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 18-260 of human PTGS2 (NP_000954) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: PTGS2
Clonality: Monoclonal
Clone: 9H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human