Anti-NKX2-1 Mouse Monoclonal Antibody [clone: 10F3]
Catalog # ORIGTA803331
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:NK2 homeobox 1
- Antigen symbol:NKX2-1
- Clonality:Monoclonal
- Clone:10F3
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human NKX2 ( NP_003308) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: NKX2-1
Clonality: Monoclonal
Clone: 10F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human