Anti-C11orf67 Mouse Monoclonal Antibody [clone: 1G1]
ORIGTA803314
: OriGene
- Antibody type:Primary
- Antigen symbol:C11orf67
- Clonality:Monoclonal
- Clone:1G1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:13.2 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human C11orf67(NP_078960) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: C11orf67
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human