Anti-ABCB1 Mouse Monoclonal Antibody [clone: 10H3]
Catalog # ORIGTA806730
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family B, member 1
- Antigen symbol:ABCB1
- Clonality:Monoclonal
- Clone:10H3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Amino acid number:995 to 1280
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 995-1280 of human ABCB1 (NP_000918) produced in SF9 cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:150
Type: Primary
Antigen: ABCB1
Clonality: Monoclonal
Clone: 10H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human