Anti-ATP5O Mouse Monoclonal Antibody [clone: 2E9]
Catalog # ORIGTA804572
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:ATP Synthase, H+ transporting, mitochondrial F1 complex, O subunit
- Antigen symbol:ATP5O
- Clonality:Monoclonal
- Clone:2E9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:24 to 213
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:20.8 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 24-213 of human ATP5O (NP_001688) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: ATP5O
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human