Anti-VWF Mouse Monoclonal Antibody [clone: 4B12]
Catalog # ORIGTA803274
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:VWF
- Clonality:Monoclonal
- Clone:4B12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:764 to 1263
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 764-1263 of human VWF (NP_000543) produced in SF9 cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human