Anti-NKX3-1 Mouse Monoclonal Antibody [clone: 2D2]
Catalog # ORIGTA805167
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:NK3 homeobox 1
- Antigen symbol:NKX3-1
- Clonality:Monoclonal
- Clone:2D2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:26.2 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human NKX3-1 (NP_006158) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: NKX3-1
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human