Anti-RIPK3 Mouse Monoclonal Antibody [clone: 1B3]
ORIGTA803100
: OriGene
- Antibody type:Primary
- Antigen name:Receptor-interacting serine-threonine kinase 3
- Antigen symbol:RIPK3
- Clonality:Monoclonal
- Clone:1B3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:233 to 518
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:56.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 233-518 of human RIPK3 (NP_006862) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150
Type: Primary
Antigen: RIPK3
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human