Anti-NR4A3 Mouse Monoclonal Antibody [clone: 1C5]
Catalog # ORIGTA804894
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:NR4A3
- Clonality:Monoclonal
- Clone:1C5
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:332 to 626
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:68 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 332-626 of human NR4A3 (NP_008912) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: NR4A3
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human