Anti-DOT1L Mouse Monoclonal Antibody [clone: 2B10]
ORIGTA802479
: OriGene
- Antibody type:Primary
- Antigen name:DOT1-Like
- Antigen symbol:DOT1L
- Clonality:Monoclonal
- Clone:2B10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:1 to 434
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1-434 of human DOT1L (NP_115871) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: DOT1L
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human