Anti-MEOX1 Mouse Monoclonal Antibody [clone: 1D1]
Catalog # ORIGTA804789
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:MEOX1
- Clonality:Monoclonal
- Clone:1D1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:27.8 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human MEOX1 (NP_004518) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: MEOX1
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human