Anti-CD20 IgG2a Monoclonal Antibody [clone: 743Y4]
Catalog # ORIGTA591082
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CD20
- Clonality:Monoclonal
- Clone:743Y4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:Concentrated
- Storage buffer:PBS with 50% glycerol and 0,05% azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:A full length cDNA gene delivered by DNA immunization
- Purification:Protein A purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Use at 0.1-1ug/ml. Flow Cytometry , CDC
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 743Y4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human