Anti-CXCL16 Rabbit Polyclonal Antibody
Catalog # ORIGTA328288
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CXCL16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:A sterile filtered antibody solution was lyophilized from PBS, pH 7,2.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:E.coli derived Recombinant Human CXCL16
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CXCL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human