Anti-ANKMY2 Mouse Monoclonal Antibody [clone: 2C3]
Catalog # ORIGTA507304
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:ankyrin repeat and MYND Domain containing 2
- Antigen symbol:ANKMY2
- Clonality:Monoclonal
- Clone:2C3
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:49.1 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human ANKMY2(NP_064715) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:400-4000
Type: Primary
Antigen: ANKMY2
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human