Anti-NFKBIA Mouse Monoclonal Antibody [clone: 1D4]
Catalog # ORIGTA507172
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, alpha
- Antigen symbol:NFKBIA
- Clonality:Monoclonal
- Clone:1D4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:35.4 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human NFKBIA(NP_065390) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100
Type: Primary
Antigen: NFKBIA
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human